Culture inserts, BRAND Insert 2in1, for 6-well plates, sterile, BIO-CERT® cell culture quality
Toimittaja: Brand
782752 782753 782754 782755
734-3683EA 367 EUR
734-3683 734-3684 734-3685 734-3686
Culture inserts, BRAND Insert 2in1, for 6-well plates, sterile, BIO-CERT® cell culture quality
Kuoppalevyt Inserttilevyt
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
  • Inserts can be used in a hanging or standing position
  • For 24, 12, and 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth thanks to surface treatment
  • Suitable for 2D and 3D cell cultures
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, savingMore Product Information