Culture inserts, BRAND Insert 2in1, for 12-well plates, sterile, BIO-CERT® cell culture quality
Toimittaja: Brand
782732 782733 782734 782735
734-3679EA 572 EUR
734-3679 734-3680 734-3681 734-3682
Culture inserts, BRAND Insert 2in1, for 12-well plates, sterile, BIO-CERT® cell culture quality
Kuoppalevyt Inserttilevyt
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
  • Inserts can be used in a hanging or standing position
  • For 24-, 12-, and 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth thanks to surface treatment
  • Suitable for 2D and 3D cell cultures
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, savingMore Product Information