VWR®, Pullot, Roller-pullot
Toimittaja: VWR Collection
734-2749EA 341 EUR
734-2749 734-2750 734-2752 734-3248 734-3249 734-3250 734-3251 734-3368 734-3369
VWR®, Pullot, Roller-pullot
Pullot Pyörivät pullot
Roller bottles are optimal vessels for large scale cell growth and multiplication, that are suitable for both research and manufacturing applications, including the growth of mammalian cells for virus propagation and bioproduction.
  • Caps feature large knurls for easy grip
  • Two different cap styles can be used in both open and closed systems
  • Every bottle features silk-printed graduations
  • Non pyrogenic
  • Temperature range: –20 to +50 °C
  • Every case features a labelled lot number
  • Sterilised by electron beam irradiation
These vessels suit automation and fitMore Product Information