VWR® 3-D scaffold, Cell Culture Dishes
Toimittaja: VWR Collection
734-2967EA 523 EUR
734-2967 734-2968 734-2969
VWR® 3-D scaffold, Cell Culture Dishes
Soluviljelyalustat Soluviljelymaljat
VWR® 3-D scaffold cell culture dishes, made of polystyrene (GPPS) with polystyrene polymer scaffold, are able to simulate the three-dimensional structure of cells in animals and the human body to maximum effect, providing an ideal environment for cell interaction, maximising culture area and improving yield.
  • Highly consistent scaffold (mean fibre diameter of 500 with 260 μm pore spacing) with 3-D channels providing high connectivity, facilitating nutrient transmission, consistent metabolic activity, and the accuracy of culture results
  • Cytokine and growth