VWR® Bottle Assemblies, PC, Sterile, Single-Use
Toimittaja: VWR Collection
734-2852EA 1140 EUR
734-2852 734-2853 734-2854 734-2855 734-2856 734-2895
VWR® Bottle Assemblies, PC, Sterile, Single-Use
Pullot Väliainepullot
Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-