HTS-mikrolevyt, 384-kuoppainen, vakiokorkeus, ShallowWell-levy, Nunc™
Toimittaja: Thermo Fisher Scientific
264705
734-2877EA 321 EUR
734-2877
HTS-mikrolevyt, 384-kuoppainen, vakiokorkeus, ShallowWell-levy, Nunc™
Kuoppalevyt Levyt suurtehoseulontaan
Nunc™ 384 ShallowWell standard height, black and white plates save costs in samples and reagents by reducing the total well volume to 25 μl. The standard height of the plate allows the use of automation without making adjustments to robotics, readers or liquid handlers.