VWR®, Pestles for Cell Strainers
Toimittaja: VWR Collection
734-2819EA 113 EUR
734-2819 734-2820
VWR®, Pestles for Cell Strainers
Huhmareet ja survimet Huhmareet
Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
  • Grooved handle for ease of use
  • Sterilised by electron beam irradiation
  • Non-pyrogenic, DNase/RNase free
Pakkaus: Each package has lot number printed on it to facilitate quality tracking. Supplied as individually packaged in easy to tear paper and plastic for easy opening (100/case). Bulk packaging has 25/bag and 50/case.