Cell culture flasks, Corning® PureCoat™ ECM mimetic, fibronectin peptide or collagen I peptide
Toimittaja: Corning
356242 356243 356272 356273 356275 356245
734-2584EA 757 EUR
734-2584 734-2585 734-2496 734-2497 734-2699 734-2697
Cell culture flasks, Corning® PureCoat™ ECM mimetic, fibronectin peptide or collagen I peptide
Kolvit ja erlenmeyerit Soluviljelypullot
Corning® PureCoat™ ECM Mimetic flasks are coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent alternative to complex extracellular peptides. The unique three or five layer Multi-Flask design lets you grow more cells in the same amount of incubator space, while the pre-coated Fibronectin or Collagen I peptide surfaces eliminate time-consuming and inconsistent self-coating protocols. The PureCoat ECM Mimetic Multi-Flasks have the same footprint, reagent volume, and cell seeding densities per unit areaMore Product Information