Cellulose Soxhlet extraction thimbles, Whatman™
Toimittaja: Whatman products (Cytiva)
2800-105 2800-185 2800-199 2800-226 2800-228 2800-250 2800-258 2800-266 2800-280 2800-282 2800-288 2800-300 2800-308 2800-330 2800-331 2800-338 2800-339 2800-373 2800-412 2800-432 2800-608 2810-228 2810-258 2810-338 2810-432 2810-339 2800-259 2800-260 2810-166 2810-266 2810-902
734-2500EA 155 EUR
734-2500 734-2501 734-2502 734-2578 734-2503 734-2504 734-2505 734-2506 734-2507 734-2508 734-2509 734-2510 734-2511 734-2512 734-2513 734-2514 734-2515 734-2516 734-2517 734-2518 734-2519 515-0167 515-0168 515-0169 515-8943 537-0036 734-2579 734-2580 537-0034 537-0035 WHAT2810-902
Cellulose Soxhlet extraction thimbles, Whatman™
Uuttolaitteet ja uutoshylsyt Uutoshylsyt
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120 °C (248 °F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1 to 1,5 mm to support application need
  • Option for double thickness for added rigidity