Roller-pullo, Nunc™ InVitro™
Toimittaja: Thermo Fisher Scientific
1060-20 1060-85 1760-20 2160-05 2160-20 4260-22 1060-05 1860-22
734-2393EA 196 EUR
734-2393 734-2394 734-2396 734-2397 734-2694 734-2695 734-2392 734-2395
Roller-pullo, Nunc™ InVitro™
Pullot Pyörivät pullot
PETG, sterile.
  • Quick-action ergonomic closure reduces wrist stain and increases productivity
  • Wide range of sizes with surface areas from 1050 to 4200 cm²
  • Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
  • Easy-to-read graduations for medium fills
  • Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.