Soluviljelylasit pulloilla, Nunc™
Toimittaja: Thermo Fisher Scientific
170920 177453
734-2107EA 682 EUR
734-2107 732-2609
Soluviljelylasit pulloilla, Nunc™
Alus- ja peitinlasit Kammiolliset aluslasit
Glass (Flaskette), PS (SlideFlask), sterile for cell culture on a microscope slide.
  • PS SlideFlask Nunclon™Δ certified
  • SlideFlask is ultrasonically welded to the slide, and individually leak tested
For karyotyping of cells, single cell autoradiography and single cell immuno-fluorescence.
W×D (Flaskette): 20×52 mm; W×D (SlideFlask): 18×50 mm