Standard polystyrene microplates 96-well, Corning® Stripwell™
Toimittaja: Corning
2580 2592 2593 3923 3924 9102 9102BC
732-5504EA 513 EUR
732-5504 734-1514 734-1515 732-5547 732-5548 732-5569 CORN9102BC
Standard polystyrene microplates 96-well, Corning® Stripwell™
Kuoppalevyt Mikrolevyt immunologiaan
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation