Round bottom high clarity polypropylene tube, disposable, Falcon®
Toimittaja: Corning
352006 352018 352053 352059 352063 352002
734-0438EA 235 EUR
734-0438 734-0985 525-0123 734-0446 734-0447 391-0000
Round bottom high clarity polypropylene tube, disposable, Falcon®
Putket Viljelyputket
These disposable, clear polypropylene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 17×100 mm tubes have blue printed graduations and white writing patch
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile,