HTS-mikrolevyt, mikrotiterlevyt, 96-kuoppaiset, Sterilin®
Toimittaja: Sterilin
612U96 612V96 612F96 611U96 611V96 611F96
634-0010EA 408 EUR
634-0010 734-0483 734-0482 634-0008 634-0009 734-0481
HTS-mikrolevyt, mikrotiterlevyt, 96-kuoppaiset, Sterilin®
Kuoppalevyt Levyt suurtehoseulontaan
PS, non treated. Used for serology and microbiology. Compatible with automated plate readers and washers.
  • Suitable for use at a wavelength of 340 nm
  • Condensation rings on the lid (supplied separately) together with raised well rims on the base help minimise the risk of contamination from surrounding wells
  • Frosted write-on area on end wall, orientation corners and alpha numeric labelling ensure easy sample identification
  • Hanging well design ensures even temperature distribution around each well
  • Manufactured from virgin polystyrene for excellent optical