Round-bottom polystyrene tubes, disposable, Falcon®
Toimittaja: Corning
352001 352003 352008 352017 352025 352027 352037 352045 352051 352052 352054 352057 352058 352235
734-0435EA 255 EUR
734-0435 734-0436 734-0000 734-0989 734-0986 734-0439 734-0440 734-0441 525-0122 734-0442 734-0443 734-0444 734-0445 734-0001
Round-bottom polystyrene tubes, disposable, Falcon®
Putket Viljelyputket
These disposable, clear polystyrene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0,1 EU/ml (Cat. No. 352008 nonsterile)