Gelatin plates, Corning® BioCoat®
Toimittaja: Corning
354652 356652
734-0251EA 103 EUR
734-0251 734-0322
Gelatin plates, Corning® BioCoat®
Kuoppalevyt Soluviljelylevyt, monikuoppaiset
Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.
  • Promotion of cell attachment and spreading
  • Culture of normal and transfected F9 teratocarcinoma cells for gene