Levyt, päällystetty kollageeni IV:llä, Corning® BioCoat®
Toimittaja: Corning
354428 354430
734-0125EA 343 EUR
734-0125 734-0127
Levyt, päällystetty kollageeni IV:llä, Corning® BioCoat®
Kuoppalevyt Soluviljelylevyt, monikuoppaiset
Type IV Collagen is a ubiquitous component in basement membranes and provides the major structural support for this matrix. When the Collagen IV meshwork is assembled, it provides a scaffold for the assembly of other basement membrane components through interactions with laminin, entactin/nidogen, and heparan sulfate proteoglycan. Collagen IV is useful as a substrate for growth of epithelial, endothelial, muscle, and nerve cells. Collagen plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation
  • Promotion of cell attachment and spreading