VWR®, Taq DNA polymerase master mix
Toimittaja: VWR Chemicals
733-1316EA 1020 EUR
733-1316 733-1314 733-1315 733-1317 733-2540 733-2542 733-2543
VWR®, Taq DNA polymerase master mix
Nukleiinihapporeagenssit End-point PCR Enzymes and Kits
VWR® Taq DNA Polymerase Master Mix is a ready to use 1,1X or 2X reaction mix. Simply add primers, template and water to carry out primer extensions and other molecular biology applications.
Tests = Reactions