VWR® Rigid PCR Plates
Toimittaja: VWR Collection
732-4396EA 57.3 EUR
732-4396 732-4397 732-4398 732-4399 732-4400 732-4401 732-4402 732-4404 732-4406 732-4407 732-4408 732-4409 732-4410 732-4411 732-4412 732-4413 732-4414 732-4415 732-4416 732-4417 732-4418 732-4421 732-4423 732-4424
VWR® Rigid PCR Plates
Kuoppalevyt PCR-levyt
VWR® Rigid PCR Plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place VWR® Rigid PCR Plates with utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Clear polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White well options are available to enhance