VWR®, Immunoassay Microplates and Strips
Toimittaja: VWR Collection
732-3726EA 0 EUR
732-3726 732-3727 732-3751 732-3752
VWR®, Immunoassay Microplates and Strips
Kuoppalevyt Mikrolevyt immunologiaan
Designed specifically for immunoassay applications.
  • Available in 96-well or strip plate format
  • Flat bottom for optimal detection
  • High quality crystal polystyrene
  • Medium binding: 100 to 200 ng IgG/cm²
  • High binding: 400 to 500 ng IgG/cm²
  • 96-well plates or 8-well strips in 12×8 frame
Highly pure polystyrene microplates designed and optimised specifically for affinity binding assays such as ELISA's. These plates are available with high binding and medium binding surfaces.
Pakkaus: Case of 50.