VWR®, Polystyrene Assay Plates, 96- and 384-Well
Toimittaja: VWR Collection
732-3720EA 185 EUR
732-3720 732-3721 732-3722 732-3723 732-3724 732-3725 732-3728 732-3729 732-3730 732-3731 732-3734 732-3735 732-3736 732-3737 732-3738 732-3739 732-3740 732-3741 732-3742 732-3743 732-3744 732-3745 732-3746 732-3747 732-3748 732-3749 732-3750
VWR®, Polystyrene Assay Plates, 96- and 384-Well
Kuoppalevyt Levyt suurtehoseulontaan
Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
  • Flat bottom ideal for microscopic applications
  • V-bottom minimising residual liquid
  • U-bottom for optimising sample/cell collection
VWR® assay plates are available in clear, white or black polystyrene, with solid or clear bottom.
The bottom of the clear bottom plates is just 0,4 mm thick and extremely flat (±0,1 mm tolerance), ensuring high measurement precision and elimination of reading errors when performing cell-based assays usingMore Product Information