PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Toimittaja: Thermo Fisher Scientific
AB-0800 AB-0800/R AB-0800/K AB-0800/W AB-0800/B AB-0800/G AB-0800/P AB-0800/Y AB-0800/L AB-0800/Y-L AB-0800/W-L AB-0800/R-L AB-0800/P-L AB-0800/G-L AB-0800/150 AB-0800/B-L
732-4888EA 160 EUR
732-4888 732-0390 732-0388 732-0391 732-0078 732-0079 732-0389 732-0392 732-0881 732-2949 732-2948 732-2947 732-2946 732-2945 ADVAAB-0800/150 732-2944
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Kuoppalevyt PCR-levyt
PP. The low profile of the plate decreases ‘dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side-wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
  • Suitable for use with automated systems
  • Maximum well volume 0,2 ml when used with adhesive and heat seals
  • Eight holes in skirt aid plate positioning and removal from thermal cycler block
  • Clear well bottom for sample visibility
  • Certified free from DNase, RNase and human genomic DNA