96-well qPCR strip tube plate, VersiPlate
Toimittaja: Thermo Fisher Scientific
AB-1800/W
732-2590EA 181 EUR
732-2590
96-well qPCR strip tube plate, VersiPlate
Kuoppalevyt PCR-levyt
VersiPlates comprise strips of eight tubes linked to each other forming the familiar 12×8 or 96-well ANSI/SBS format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
  • White colouration improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the