Pipetinkärkiä, geelinlatauskärjet, ART™
Toimittaja: Thermo Fisher Scientific
2155 2155P 3621
732-2226EA 95.4 EUR
732-2226 732-2229 732-2255
Pipetinkärkiä, geelinlatauskärjet, ART™
Pipetinkärjet
These tips are designed for loading vertical acrylamide or agarose gels.
  • Sterile tips are certified free from RNase, DNase, ATP and pyrogen
  • Available with or without filter for trapping potential contaminants
  • Manufactured according to the industry's highest quality standard