VWR®, PCR-putkien ja korkkien nauhat
Toimittaja: VWR Collection
732-0546EA 251 EUR
732-0546 732-0550 732-0552 732-0553 732-0554 732-0545 211-0338 211-0339 211-0381 732-1506 732-1521 732-1517 732-1518 732-1519 732-1520 731-0367 732-3391 732-3484 732-3485 732-3499 732-3608 732-3609 732-3610
VWR®, PCR-putkien ja korkkien nauhat
Putket PCR-putket
These PCR tube strips and caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.