Sequencing septa mats, Axygen®
Toimittaja: Corning
AM96SEPTA3100
731-4130EA 163.78 EUR
731-4130
Sequencing septa mats, Axygen®
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.