Adhesive sealing film for microplates, QuickSeal Micro™
Toimittaja: iST Scientific Ltd
IST-125-080TS IST-125-080SS IST-125-080SR IST-125-080LS IST-125-080LR
731-0628EA 20.4 EUR
731-0628 731-0627 731-0625 731-0626 731-0624
Adhesive sealing film for microplates, QuickSeal Micro™
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Transparent peelable film for storage. Suitable for aqueous sample storage.
  • Non pierceable with average strength
  • Seal integrity range: −20 to +80 °C