Heat sealing foil, PierceASeal Foil™
Toimittaja: iST Scientific Ltd
IST-106-078LR IST-106-078LS IST-106-078SR IST-106-078SS IST-106-078TR IST-106-078TS IST-106-115LR IST-106-115SR IST-106-115TR IST-106-200LR
731-0565EA 600 EUR
731-0565 731-0569 731-0566 731-0570 731-0571 731-0573 731-0567 731-0568 731-0572 ISTSIST-106-200LR
Heat sealing foil, PierceASeal Foil™
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Pierceable foil seal with good solvent resistance. Suitable for PCR, compound storage, and sample shipping.
  • Non peelable
  • Multiple sealing / resealing permissible
  • Easy sealing surface identification
  • Seal integrity range: −20 to 120 °C
  • Pierceable