Heat sealing foil, ClearASeal Weld™
Toimittaja: iST Scientific Ltd
IST-102-078TS IST-102-078LR IST-102-078LS IST-102-078SR IST-102-078TR IST-102-115LR IST-102-115SR IST-102-115TR IST-102-078SS
731-0546EA 20.4 EUR
731-0546 731-0493 731-0497 731-0494 731-0499 731-0495 731-0496 731-0545 731-0498
Heat sealing foil, ClearASeal Weld™
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Clear strong bonding film ideal for water thermal cyclers. Suitable for qPCR, PCR (water bath thermal cycling), storage, sample inspection, disposal of hazardous materials, use with DMSO.
  • Non pierceable and non peelable
  • Permanent seal (for PP plates) with solvent resistence
  • Good optical optical clarity
  • Seal integrity range: −80 to +110 °C