Heat sealing foil, ClearASeal Pierce™
Toimittaja: iST Scientific Ltd
IST-103-078TS IST-103-115SR IST-103-078LR IST-103-078LS IST-103-078SR IST-103-078SS IST-103-078TR IST-103-115LR IST-103-115TR
731-0492EA 20.4 EUR
731-0492 731-0487 731-0484 731-0488 731-0485 731-0489 731-0490 731-0486 731-0491
Heat sealing foil, ClearASeal Pierce™
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Clear pierceable film ideal for PP, PE, PS and COC plates. Suitable for use with the ABI 3730 sequencer as the thinner structure pierces more easily.
  • Non peelable
  • Moderate solvent resistance with good optical clarity
  • Seal integrity range: −80 to +80 °C (or 110 °C with pressurised PCR heated lids)