PCR microtubes with caps, 0,2 ml and 0,5 ml, Amplitube™
Toimittaja: Simport Scientific
T325-3B T325-3G T325-3N T325-3R T325-3Y T325-1B T325-1G T325-1N T325-1R T325-1Y T325-2N T325-2B T325-2G T325-2R T325-2Y T325-4N T325-4B T325-4G T325-4R T325-4Y T325-2VG T325-1VB T325-1VG T325-1VN T325-1VR T325-1VY T325-2VB T325-2VN T325-2VR T325-2VY
211-3312EA 63.5 EUR
211-3312 211-3313 211-3311 211-3314 211-3315 211-3302 211-3303 211-3301 211-3304 211-3305 211-3306 211-3307 211-3308 211-3309 211-3310 211-3316 211-3317 211-3318 211-3319 211-3320 731-0470 731-0464 731-0465 731-0466 731-0467 731-0468 731-0469 731-0471 731-0472 731-0473
PCR microtubes with caps, 0,2 ml and 0,5 ml, Amplitube™
Putket PCR-putket
PP. Ultra-thin wall design ensures rapid thermal transfer and a reduction in cycle and PCR reaction time.
  • Attached hinged caps provide positive sealing during thermal cycling, prevent evaporation and are easy to open and close with one hand
  • Choice of natural and four non-cytotoxic and non-metallic colours for visual coding of samples
  • 0,2 ml tube has a frosted writing surface and has a cap with an integral shield, which prevents contamination with the lid surface
  • 0,5 ml tube has an etched writing surface and is graduated in 0,1 ml increments
  • Certified