Adhesive PCR film seals, QuickSeal PCR™
Toimittaja: iST Scientific Ltd
IST-120-080LR IST-120-080SR IST-120-080LS IST-120-080SS IST-120-080TS
731-0462EA 1300 EUR
731-0462 731-0448 731-0447 731-0449 731-0450
Adhesive PCR film seals, QuickSeal PCR™
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Transparent, peelable film for PCR.
  • Non pierceable
  • Available sterile or non sterile
  • Seal integrity range: −20 to +110 °C