Electroporation system, AgilePulse™ in vivo

Toimittaja: BTX
47-0400N 47-0500N
731-0341EA 25590 EUR
731-0341 731-0360
Electroporation system, AgilePulse™ in vivo
Elektroporaatio Elektroporaatiolaitteet
Effectively introduced DNA vaccines represent a powerful and safe means for stimulating an immune response that recognises and eliminates target molecules in the body. However, traditional DNA vaccine delivery systems suffer from poor efficiency. The BTX AgilePulse™ in vivo system used for vaccine development and gene therapy provides an intra-dermal/intra-muscular electroporation solution to produce maximum transfection efficiency. The AgilePulse™ in vivo system can be purchased with software supporting intra-dermal (ID) or intra-muscular (IM) applications.

For vaccine applications, DNAMore Product Information
Order Now

TEKNISET TIEDOT

L×S×K 320×200×400 mm
Environmentally Preferable
Liitännät USB flash key
Jännitealue 50 - 1000 V
Pulssin leveys 0,050 to 10 ms
Pulssiväli 0,200 - 1000 ms (5 kHz - 1 Hz)
Näyttö Touch screen
Paino 11,3 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR