8 PCR-putken liuskat, irralliset tulppaliuskat
Toimittaja: Brand
781326 781327
731-0208EA 375 EUR
731-0208 731-0209
8 PCR-putken liuskat, irralliset tulppaliuskat
Putket PCR-putket
PP. Eight connected 0,2 ml tubes and separate domed or flat caps in strips of 8.
  • Flat caps are suitable for real-time PCR (qPCR)
  • Easy to open and close without tools
  • The tubes have a holding strap at one end, and the cover strips have a small lip on the side of each cap for careful, contamination-free opening
  • DNase-, RNase- and DNA-free
Colour: Clear