DNA specimen collection cards, Ahlstrom-Munksjö GenReleaz™
Toimittaja: Ahlstrom-Munksjö
8.295.0001.B-N 8.295.0000.B-N 8.295.0002.B-N 8.296.0001.B-N 8.296.0000.B-N 8.296.0002.B-N
730-0623EA 640 EUR
730-0623 730-0530 730-0625 730-0654 730-0627 730-0656
DNA specimen collection cards, Ahlstrom-Munksjö GenReleaz™
Nukleiinihapon säilytysjärjestelmät
GenReleaz™ and GenReleaz™ Colour specimen collection cards are specifically developed for collection, transport, preservation and fast and efficient elution of DNA from biological fluids stored at ambient temperature.
  • Easy sample collection
  • Stabilisation of DNA inside fiber-based material
  • Long-term ambient preservation of DNA
  • Fast and simple DNA elution
  • Less than 30 minutes processing time
  • Cost saving with no need of cold-chains
One of the key features of GenReleaz™ is the method for recovering DNA: efficient elution process eliminates the need to useMore Product Information