2-Propanoli, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

72299-1L 72299-2.5L 72299-4X2.5L 72299-6X1L
72299-1LEA 46.4 EUR
72299-1L 72299-2.5L HONC72299-4X2.5L HONC72299-6X1L
2-Propanoli, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
2-Propanoli

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635639

Kaava: (CH₃)₂CHOH
Kiehumispiste: 77…83 °C (1,013 hPa)
Sulamispiste: –90 °C
Tiheys: 0,784…0.786 g/cm³ (20 °C)
Leimahduspiste: 12 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011674
CAS-numero: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance complying
Assay (GC) Min. 99.9 %
IR spectrum complying
Boiling range 82.1 - 82.6 °C
Density (D 20/4) 0.784 - 0.786
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 10 ppm
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₂H₅COOH) Max. 0.00074 %
Free alkali (as NH₃) Max. 0.00017 %
Free acid, free alkali Max. 0.0001 meq/g
Peroxides (as H₂O₂) Max. 0.00034 %
Reaction against H₂SO₄ complying
KMnO₄ reducing matter (as O) Max. 5 ppm
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.01 ppm
Heavy metals (as Pb) Max. 1 ppm
Carbonyl compounds (as Acetone) Max. 20 ppm
Carbonyl compounds (as propionaldehyde) Max. 20 ppm
Ethanol (GC) Max. 0.01 %
Methanol (GC) Max. 0.1 %
APHA Max. 10
Solubility in water complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR