SUPERFLO™ Histology Cassettes, Hinged Design
Toimittaja: Simport Scientific
M533-10SL M533-11 M533-11SL M533-12 M533-12SL M533-2 M433-6T M433-7T M433-8T M433-9T M434-5T M434-6T M434-7T M434-8T M434-9T M435-10T M435-11T M435-12T M435-2T M435-4T M435-5T M435-6T M435-7T M435-8T M435-9T M433-10T M433-11T M433-12T M433-2T M433-3T M433-4T M433-5T M434-10T M434-11T M434-12T M434-2T M434-3T M434-4T M533-9SL M534-10 M534-10SL M534-11 M435-3T M534-3SL M534-4 M534-4SL M534-5 M534-5SL M534-6 M534-6SL M534-7 M534-7SL M534-8SL M534-9 M534-9SL M535-10 M535-10SL M535-11 M533-10 M533-2SL M533-3 M533-3SL M533-4 M533-4SL M533-5 M533-5SL M533-6 M533-6SL M533-7 M533-7SL M533-8 M533-8SL M533-9 M534-11SL M534-12 M534-12SL M534-2 M534-2SL M534-3 M535-7SL M535-8 M535-8SL M535-9 M534-8 M535-11SL M535-12 M535-12SL M535-2 M535-2SL M535-3 M535-3SL M535-4 M535-4SL M535-5 M535-5SL M535-6 M535-6SL M535-7 M535-9SL
720-3002EA 28.6 EUR
720-3002 720-3001 720-3000 720-2999 720-2998 720-2997 SIMPM433-6T SIMPM433-7T 720-3005 720-3004 SIMPM434-5T SIMPM434-6T SIMPM434-7T SIMPM434-8T SIMPM434-9T SIMPM435-10T SIMPM435-11T SIMPM435-12T SIMPM435-2T SIMPM435-4T SIMPM435-5T SIMPM435-6T SIMPM435-7T SIMPM435-8T SIMPM435-9T SIMPM433-10T SIMPM433-11T SIMPM433-12T SIMPM433-2T SIMPM433-3T SIMPM433-4T SIMPM433-5T SIMPM434-10T SIMPM434-11T SIMPM434-12T SIMPM434-2T SIMPM434-3T SIMPM434-4T 720-2982 SIMPM534-10 SIMPM534-10SL SIMPM534-11 SIMPM435-3T SIMPM534-3SL SIMPM534-4 SIMPM534-4SL SIMPM534-5 SIMPM534-5SL SIMPM534-6 SIMPM534-6SL SIMPM534-7 SIMPM534-7SL SIMPM534-8SL SIMPM534-9 SIMPM534-9SL SIMPM535-10 SIMPM535-10SL SIMPM535-11 720-3003 720-2996 720-2995 720-2994 720-2993 720-2992 720-2991 720-2990 720-2989 720-2988 720-2987 720-2986 720-2985 720-2984 720-2983 SIMPM534-11SL SIMPM534-12 SIMPM534-12SL SIMPM534-2 SIMPM534-2SL SIMPM534-3 SIMPM535-7SL SIMPM535-8 SIMPM535-8SL SIMPM535-9 SIMPM534-8 SIMPM535-11SL SIMPM535-12 SIMPM535-12SL SIMPM535-2 SIMPM535-2SL SIMPM535-3 SIMPM535-3SL SIMPM535-4 SIMPM535-4SL SIMPM535-5 SIMPM535-5SL SIMPM535-6 SIMPM535-6SL SIMPM535-7 SIMPM535-9SL
SUPERFLO™ Histology Cassettes, Hinged Design
Histologiset kasetit Kasetit kudosten käsittelyyn
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface.
  • Unique lateral vents, fluids can access tissue from all four sides
  • Basket style for small tissue management
  • Special hinged cover design with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colours
Molded from high-density polymer, keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-More Product Information