Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Toimittaja: Simport Scientific
M480-10T M480-11T M480-12T M480-2T M480-3T M480-4T M480-5T M480-6T M480-7T M480-8T M480-9T M480-10SL M480-11SL M480-12SL M480-2SL M480-3SL M480-4SL M480-5SL M480-6SL M480-7SL M480-8SL M480-9SL
720-1923EA 511 EUR
720-1923 720-1925 720-1927 720-1929 720-1931 720-1933 720-1935 720-1937 720-1939 720-1941 720-1943 720-1922 720-1924 720-1926 720-1928 720-1930 720-1932 720-1934 720-1936 720-1938 720-1940 720-1942
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Histologiset kasetit Kasetit kudosten käsittelyyn
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
  • The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
  • Keep specimens safely submerged
  • Totally resistant to the chemical action of solvents used in histology laboratories