Base moulds, stainless steel
Toimittaja: Simport Scientific
M474-1 M474-3 M474-2 M474-4 M474-5
720-1917EA 140 EUR
720-1917 720-1919 720-1918 720-1920 720-1921
Base moulds, stainless steel
Muotit Moulds for Histology
Made of stainless steel. Sizes fit most cassettes. Superior thermal exchange due to rounded corners. These moulds are for all applications in specimen embedding with Simport embedding rings and cassettes. Manufactured from high quality stainless steel for optimal thermal conductivity.
  • The moulds have a highly polished surface for easy paraffin block removal
  • Well corners are rounded for optimal paraffin ribboning