Tissue processing/embedding cassettes, Swingsette®, M515
Toimittaja: Simport Scientific
M515-10 M515-11 M515-12 M515-4 M515-5 M515-2 M515-3 M515-6 M515-7 M515-8 M515-9
720-1628EA 251 EUR
720-1628 720-1629 720-1630 720-1636 720-1637 720-1634 720-1635 720-1638 720-1639 720-1640 720-1641
Tissue processing/embedding cassettes, Swingsette®, M515
Histologiset kasetit Kasetit kudosten käsittelyyn
Asetaalipolymeeristä
  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.
Pakkaus: Each case contains three boxes of 500 cassettes.