Biopsy processing/embedding cassettes, Unisette™ M506
Toimittaja: Simport Scientific
M506-2 M506-3 M506-4 M506-5 M506-6 M506-7 M506-8 M506-9 M506-10 M506-11 M506-12
720-1570EA 260 EUR
720-1570 720-1571 720-1572 720-1573 720-1574 720-1575 720-1576 720-1577 720-1564 720-1565 720-1566
Biopsy processing/embedding cassettes, Unisette™ M506
Histologiset kasetit Kudos- ja biopsiakasetit
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Integral lid
  • Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
  • Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
Pakkaus: Each case contains three boxes of 500 cassettes each.