Biopsiakasetit QuickLoad™-telineissä, Slimsette™, M510T
Toimittaja: Simport Scientific
M510-10T M510-11T M510-12T M510-2T M510-3T M510-4T M510-5T M510-6T M510-7T M510-8T M510-9T
720-1447EA 867 EUR
720-1447 720-1449 720-1451 720-1456 720-1458 720-1460 720-1462 720-1464 720-1466 720-1468 720-1470
Biopsiakasetit QuickLoad™-telineissä, Slimsette™, M510T
Histologiset kasetit Kudos- ja biopsiakasetit
Made of acetal. Slimsette™ biopsy cassettes are similar to M509 tissue cassettes but are specially designed to hold biopsy specimens.
  • The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface
  • Available in several colours
They have an attached lid that opens from the back of the cassette. The lids arrive closed but can be opened many times, always relocking securely.
Pakkaus: Cassettes are packed in stacks of 40. Each case contains 50 stacks for a total of 2000 cassettes.