Biopsiakasetit QuickLoad™-koteloissa, Slimsette™, M510SL
Toimittaja: Simport Scientific
M510-10SL M510-11SL M510-12SL M510-2SL M510-3SL M510-4SL M510-5SL M510-6SL M510-7SL M510-8SL M510-9SL
720-1446EA 323 EUR
720-1446 720-1448 720-1450 720-1455 720-1457 720-1459 720-1461 720-1463 720-1465 720-1467 720-1469
Biopsiakasetit QuickLoad™-koteloissa, Slimsette™, M510SL
Histologiset kasetit Kudos- ja biopsiakasetit
Acetal polymer.
  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems
Pakkaus: Cassettes are packed in sleeves of 75. Each case contains 10 sleeves.