Staining dishes, WHEATON®
Toimittaja: DWK Life Sciences
900403 900400 900401 900402 900404
631-2506EA 1190 EUR
631-2506 720-1031 720-1032 720-1033 631-2507
Staining dishes, WHEATON®
Astiat ja purkit Värjäystölkki
Soda-lime glass and stainless steel.
  • Handle permanently attached
  • Hinged closure
  • Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.