Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Tonbo Biosciences
72-8080-M001
72-8080-M001EA 215 EUR
72-8080-M001
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Donkey anti-Goat IgG (H&L) - Affinity Pure, HRP Conjugate, min x w/human, mouse, rabbit or rat IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR