J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Toimittaja: Avantor
7182-02EA 0 EUR
7182-02 7182-05 7285-00 7285-01
J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Proteiininpuhdistusjärjestelmät Protein Purification Resins
Silica-based media used for the purification of proteins based on differential salt induced hydrophobicity. Mixed mode provides unique selectivity with its hydrophobic interaction and ion exchange function.