D-Desthiobiotin
Toimittaja: Merck Millipore (Novagen)
71610-3
71610-3EA 805 EUR
71610-3
D-Desthiobiotin
Proteiininpuhdistusjärjestelmät Protein Purification Resins
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.