Cuvette tests for water testing for WTW spectrophotometers and photometers
Toimittaja: WTW
250303 250309 250313 250314 250341 250344 250349 250358 252075 250406 250408 250409 250410 250411 250412 250413 250414 250415 250416 250417 250436 250450 250304 250329 250347 250351 250359 250361 250402 250403 250404 250407 250424 250494 250495 252023 250420 252039 252018 252061 252073 252074 252071 252072 252070 252076 252028 250421 252062 252036 252045 252051 252040 252066 252029 252024 252025 252091
713-4007EA 109 EUR
713-4007 713-4013 713-4004 713-4005 713-4006 WTWA250344 WTWA250349 WTWA250358 713-4032 WTWA250406 713-4023 713-4024 713-4027 713-4028 713-4031 713-4036 713-2237 713-4041 WTWA250416 713-4045 WTWA250436 WTWA250450 713-4010 713-4002 713-4029 713-4012 713-4009 713-4017 713-4044 713-4039 713-4021 713-4022 713-4003 713-4038 713-4000 WTWA252023 WTWA250420 WTWA252039 WTWA252018 WTWA252061 WTWA252073 WTWA252074 WTWA252071 WTWA252072 WTWA252070 WTWA252076 WTWA252028 WTWA250421 WTWA252062 WTWA252036 WTWA252045 WTWA252051 WTWA252040 705-0725 WTWA252029 WTWA252024 WTWA252025 WTWA252091
Cuvette tests for water testing for WTW spectrophotometers and photometers
Ympäristöanalytiikan pikatestit
A wide choice of tests is available, for routine quantitative analysis in different applications, designed for use with WTW spectrophotometers and photometers. The test is automatically recognised by the laboratory instruments (via a barcoding system) and results are displayed in the appropriate units. Due to the different photometer optics, one test may yield different measuring ranges in different instruments; LED photometers may have smaller measuring ranges for the same test.