PCR-putkiliuskat ja Masterclear®-tulppaliuskat
Toimittaja: EPPENDORF
0030132.890 0030132.882 0030132.874
713-0018EA 309 EUR
713-0018 713-0017 713-0016
PCR-putkiliuskat ja Masterclear®-tulppaliuskat
Putket PCR-putket
Real-time PCR tube strips feature an extremely thin wall for optimal heat transfer. Designed to provide high mechanical stability, they are ideal for real-time PCR applications, especially for use with small reaction volumes. White wells offer improved reflection.
Masterclear™ cap strips offer distinct advantages for real-time PCR applications. The inverted dome of the cap strips prevents scratching or contamination of the optical surface and reduces the volume of the micro test tube. Additionally, the material and the production processes are optimised to ensure excellent light transmission.