easySpiral® spiral plater
Toimittaja: INTERSCIENCE
412000 413000
710-2258EA 0 EUR
710-2258 710-2259
easySpiral® spiral plater
Kierrelevittimet
easySpiral® and easySpiral® Pro spiral platers, two very fast platers. A predetermined volume of bacterial suspension is sprayed in the shape of a spiral in a logarithmic dilution onto four areas of the Petri dish. The deposited volume corresponds to the set dilution factor. With easySpiral® Pro, it is possible to export to Excel.
  • Improved technology for spiral shaped plating of bacteria
  • A newly designed rotating arm allows for a complete cycle in just 25 seconds!
  • The Spiral® method reduces the dilution steps by ¼
Counting range: 100 to 1×10⁷ CFU/ml (More Product Information